missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(3-hydroxyphenoxy)benzene 96.0+%, TCI America™

Catalog No. B14851G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B14851G Supplier TCI America Supplier No. B14851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5085-95-0
Molecular Formula C18H14O4
Molecular Weight (g/mol) 294.31
MDL Number MFCD00142543
InChI Key CHBWEVDVYGBMEJ-UHFFFAOYSA-N
Synonym Hydroquinone Bis(3-hydroxyphenyl) Ether
PubChem CID 626369
IUPAC Name 3-[4-(3-hydroxyphenoxy)phenoxy]phenol
SMILES OC1=CC(OC2=CC=C(OC3=CC=CC(O)=C3)C=C2)=CC=C1

Specifications

Melting Point 126°C
Quantity 1 g
Formula Weight 294.31
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(3-hydroxyphenoxy)benzene