missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene, 98%, Thermo Scientific Chemicals

Catalog No. AAB2025903
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2025903 1 g
AAB2025909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2025903 Supplier Thermo Scientific Chemicals Supplier No. B2025903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1992-15-0
Molecular Formula C12H6F12O2
Molecular Weight (g/mol) 410.16
MDL Number MFCD00042091
InChI Key YTJDSANDEZLYOU-UHFFFAOYSA-N
Synonym 1,4-bis hexafluoro-alpha-hydroxyisopropyl benzene, 2,2'-1,4-phenylene bis 1,1,1,3,3,3-hexafluoropropan-2-ol, 1,4-bis 2-hydroxyhexafluoroisopropyl benzene, 1,4-bis perfluoro 2-hydroxyprop-2-yl benzene, 1,1,1,3,3,3-hexafluoro-2-4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,4-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,4-benzenedimethanol, 1,1,1,3,3,3-hexafluoro-2-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethy l phenyl propan-2-ol, 2,2'-p-phenylene bis 1,1,1,3,3,3-hexafluoro-2-propanol, 1,4-bis 2-hydroxyhexafluoro-2-propyl benzene
PubChem CID 631065
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
SMILES OC(C1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 92°C to 95°C
Boiling Point 100°C (20 mmHg)
Quantity 1 g
Formula Weight 410.16
Percent Purity 98%
Chemical Name or Material 1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only