missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(2-hydroxyethoxy)benzene 95.0+%, TCI America™

Catalog No. p-7126578
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D106525G 25 g
D1065500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D106525G Supplier TCI America Supplier No. D106525G
Only null left

Chemical Identifiers

CAS 104-38-1
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
MDL Number MFCD00002861
InChI Key WTPYFJNYAMXZJG-UHFFFAOYSA-N
Synonym hydroquinone bis 2-hydroxyethyl ether, 1,4-bis 2-hydroxyethoxy benzene, vernatzer 30/10, 2,2'-1,4-phenylenebis oxy diethanol, hydroquinone di 2-hydroxyethyl ether, hydroquinone diethylol ether, 2,2'-phenylenedioxy diethanol, 2,2'-p-phenylenedioxy diethanol, 2,2'-p-phenylenedioxydiethanol, 2-4-2-hydroxyethoxy phenoxy ethanol
PubChem CID 66912
IUPAC Name 2-[4-(2-hydroxyethoxy)phenoxy]ethanol
SMILES C1=CC(=CC=C1OCCO)OCCO

Specifications

Melting Point 105°C
Quantity 25 g
Formula Weight 198.22
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(2-hydroxyethoxy)benzene
EINECSNumber (3)-0751
TSCA Yes