missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(2-benzoxazolyl)naphthalene 98.0+%, TCI America™

Catalog No. p-7119741
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B272825G 25 g
B2728500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B272825G Supplier TCI America Supplier No. B272825G

Chemical Identifiers

CAS 5089-22-5
Molecular Formula C24H14N2O2
Molecular Weight (g/mol) 362.39
MDL Number MFCD00357168
InChI Key WFYSPVCBIJCZPX-UHFFFAOYSA-N
PubChem CID 78769
IUPAC Name 2-[4-(1,3-benzoxazol-2-yl)naphthalen-1-yl]-1,3-benzoxazole
SMILES O1C2=CC=CC=C2N=C1C1=C2C=CC=CC2=C(C=C1)C1=NC2=CC=CC=C2O1

Specifications

Melting Point 212°C
Quantity 25 g
Formula Weight 362.39
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(2-benzoxazolyl)naphthalene