missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Benzoquinone 98.0+%, TCI America™
Supplier: TCI America B0089500G
Specifications
| 1,4-Benzoquinone | |
| 113°C | |
| C6H4O2 | |
| 500 g | |
| p-benzoquinone, benzoquinone, quinone, 1,4-benzoquinone, p-quinone, chinone, 2,5-cyclohexadiene-1,4-dione, cyclohexadienedione, para-quinone, 1,4-benzoquine | |
| C1=CC(=O)C=CC1=O | |
| 108.096 | |
| CHEBI:16509 | |
| ≥98.0% (T) |
| 106-51-4 | |
| Yellow | |
| MFCD00001591 | |
| 2587 | |
| AZQWKYJCGOJGHM-UHFFFAOYSA-N | |
| cyclohexa-2,5-diene-1,4-dione | |
| 4650 | |
| 108.10 | |
| Crystalline Powder |
Chemical Identifiers
| 106-51-4 | |
| 108.096 | |
| AZQWKYJCGOJGHM-UHFFFAOYSA-N | |
| 4650 | |
| cyclohexa-2,5-diene-1,4-dione |
| C6H4O2 | |
| MFCD00001591 | |
| p-benzoquinone, benzoquinone, quinone, 1,4-benzoquinone, p-quinone, chinone, 2,5-cyclohexadiene-1,4-dione, cyclohexadienedione, para-quinone, 1,4-benzoquine | |
| CHEBI:16509 | |
| C1=CC(=O)C=CC1=O |
Safety and Handling
EINECSNumber : (3)-1001
RTECSNumber : DK2625000
TSCA : Yes