missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzoquinone 98.0+%, TCI America™

Catalog No. p-7118345
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B008925G 25 g
B0089100G 100 g
B0089500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B008925G Supplier TCI America Supplier No. B008925G
Only null left

Chemical Identifiers

CAS 106-51-4
Molecular Formula C6H4O2
Molecular Weight (g/mol) 108.096
MDL Number MFCD00001591
InChI Key AZQWKYJCGOJGHM-UHFFFAOYSA-N
Synonym p-benzoquinone, benzoquinone, quinone, 1,4-benzoquinone, p-quinone, chinone, 2,5-cyclohexadiene-1,4-dione, cyclohexadienedione, para-quinone, 1,4-benzoquine
PubChem CID 4650
ChEBI CHEBI:16509
IUPAC Name cyclohexa-2,5-diene-1,4-dione
SMILES C1=CC(=O)C=CC1=O

Specifications

Melting Point 113°C
Color Yellow
Quantity 25 g
UN Number 2587
Formula Weight 108.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Benzoquinone
EINECSNumber (3)-1001
RTECSNumber DK2625000
TSCA Yes