missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxane, 98%

Catalog No. p-7023722
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1755809 10 g
AAA1755818 50 g
AAA1755830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1755809 Supplier Thermo Scientific Chemicals Supplier No. A1755809
Only null left

CAS: 493-09-4 | C8H8O2 | 136.15 g/mol

Benzo-1,4-dioxane (2,3-dihydro-1,4-benzodioxin) is used in the synthesis of stereoisomers which were evaluated as α- and β- adrenergic antagonists

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo-1,4-dioxane (2,3-dihydro-1,4-benzodioxin) is used in the synthesis of stereoisomers which were evaluated as α- and β- adrenergic antagonists

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 493-09-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00006821
InChI Key BNBQRQQYDMDJAH-UHFFFAOYSA-N
Synonym 1,4-benzodioxane, 1,4-benzodioxan, 2,3-dihydrobenzo b 1,4 dioxine, 1,4-benzodioxin, 2,3-dihydro, pyrocatechol ethylene ether, 1,2-ethylenedioxy benzene, benzodioxan, ethylene o-phenylene dioxide, 1,2-ethylenedioxybenzene, 1, 4-benzdioxan
PubChem CID 10301
IUPAC Name 2,3-dihydro-1,4-benzodioxine
SMILES C1COC2=CC=CC=C2O1

Specifications

Density 1.17
Boiling Point 216°C
Flash Point 87°C (188°F)
Refractive Index 1.549
Quantity 10 g
Beilstein 120846
Solubility Information Slightly soluble in water.
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 1,4-Benzodioxane
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 207-775-6
RTECSNumber DF4728000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only