accessibility menu, dialog, popup

UserName

CAS RN 493-09-4

IUPAC Name: 2,3-dihydro-1,4-benzodioxine
Synonyms: benzodioxan
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: BNBQRQQYDMDJAH-UHFFFAOYSA-N
SMILES: C1COC2=CC=CC=C2O1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)