missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxane-6-boronic acid, 97%

Catalog No. AAL2029606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2029606 5 g
AAL2029603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2029606 Supplier Thermo Scientific Chemicals Supplier No. L2029606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 164014-95-3 | C8H9BO4 | 179.966 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 164014-95-3
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.966
MDL Number MFCD01009696
InChI Key SQDUGGGBJXULJR-UHFFFAOYSA-N
Synonym 1,4-benzodioxane-6-boronic acid, 2,3-dihydrobenzo b 1,4 dioxin-6-yl boronic acid, 3,4-ethylenedioxy benzeneboronic acid, 2,3-dihydro-1,4-benzodioxin-6-ylboranediol, boronic acid, 2,3-dihydro-1,4-benzodioxin-6-yl, 1,4-benzodioxan-6-boronic acid, 2,3-dihydro-1,4-benzodioxin-6-yl boronic acid, 2,3-dihydrobenzo b 1,4 dioxin-6-ylboronic acid, 1,4-benzodioxan-6-yl boronic acid
PubChem CID 2776178
IUPAC Name 2,3-dihydro-1,4-benzodioxin-6-ylboronic acid
SMILES B(C1=CC2=C(C=C1)OCCO2)(O)O

Specifications

Quantity 5 g
Beilstein 7478648
Formula Weight 179.97
Percent Purity 97%
Chemical Name or Material 1,4-Benzodioxane-6-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only