accessibility menu, dialog, popup

UserName

CAS RN 164014-95-3

IUPAC Name: (2,3-dihydro-1,4-benzodioxin-6-yl)boronic acid
Synonyms: 2,3-dihydro-1,4-benzodioxin-6-ylboronic acid
Molecular Weight (g/mol): 179.97
Molecular Formula: C8H9BO4
InChi Key: SQDUGGGBJXULJR-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C2OCCOC2=C1
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)