missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxan-5-amine, 95%

Catalog No. AC429370050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429370050 5 g Glass bottle
AC429370010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC429370050 Supplier Thermo Scientific Chemicals Supplier No. 429370050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16081-45-1 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16081-45-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD03695459
InChI Key DMLRSJNZORFCBD-UHFFFAOYSA-N
Synonym 5-amino-1,4-benzodioxane, 2,3-dihydrobenzo b 1,4 dioxin-5-amine, 2,3-dihydro-benzo 1,4 dioxin-5-ylamine, 2,3-ethylenedioxyaniline, 1,4-benzodioxin-5-amine,2,3-dihydro, 1,4-benzodioxan-5-amine, 5-amino-1,4-benzodioxan, 5-amino-1,4-dibenzodioxane, 1,4-benzodioxin-5-amine, 2,3-dihydro, 5-amino-2,3-dihydro-1,4-benzodioxin
PubChem CID 11788387
IUPAC Name 2,3-dihydro-1,4-benzodioxin-5-amine
SMILES C1COC2=C(C=CC=C2O1)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Amber
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 151.17
Percent Purity 95%
Physical Form Beads
Chemical Name or Material 1, 4-Benzodioxan-5-amine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre

RUO – Research Use Only