CAS RN 16081-45-1
IUPAC Name:
2,3-dihydro-1,4-benzodioxin-5-amine
Synonyms:
2,3-dihydro-1,4-benzodioxin-5-amine
Molecular Weight (g/mol):
151.17
Molecular Formula:
C8H9NO2
InChi Key:
DMLRSJNZORFCBD-UHFFFAOYSA-N
SMILES:
NC1=C2OCCOC2=CC=C1
1
–
1
of
1
results
1
Promotions Available
1,4-Benzodioxan-5-amine, 95%
1,4-Benzodioxan-5-amine, 95%
CAS: 16081-45-1 Molecular Formula: C8H9NO2 Molecular Weight (g/mol): 151.17 MDL Number: MFCD03695459 InChI Key: DMLRSJNZORFCBD-UHFFFAOYSA-N Synonym: 5-amino-1,4-benzodioxane,2,3-dihydrobenzo b 1,4 dioxin-5-amine,2,3-dihydro-benzo 1,4 dioxin-5-ylamine,2,3-ethylenedioxyaniline,1,4-benzodioxin-5-amine,2,3-dihydro,1,4-benzodioxan-5-amine,5-amino-1,4-benzodioxan,5-amino-1,4-dibenzodioxane,1,4-benzodioxin-5-amine, 2,3-dihydro,5-amino-2,3-dihydro-1,4-benzodioxin PubChem CID: 11788387 IUPAC Name: 2,3-dihydro-1,4-benzodioxin-5-amine SMILES: C1COC2=C(C=CC=C2O1)N