missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenedithiol 98.0+%, TCI America™

$286.85 - $850.92

Chemical Identifiers

CAS 624-39-5
Molecular Formula C6H6S2
Molecular Weight (g/mol) 142.23
MDL Number MFCD00142541
InChI Key WYLQRHZSKIDFEP-UHFFFAOYSA-N
Synonym 1,4-benzenedithiol, dithiohydroquinone, 1,4-dimercaptobenzene, 4-mercaptothiophenol, 1,4-benzene dithiol, pubchem6866, acmc-1b45q, benzene-1,4-dithiol gc
PubChem CID 4691729
IUPAC Name benzene-1,4-dithiol
SMILES SC1=CC=C(S)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B15051G
View Documents
TCI America
B15051G
1 g
Each for $286.85
Only null left
 
B15055G
View Documents
TCI America
B15055G
5 g
Each for $850.92
Only null left
 
Description

Description

Specifications

Chemical Identifiers

624-39-5
142.23
WYLQRHZSKIDFEP-UHFFFAOYSA-N
4691729
SC1=CC=C(S)C=C1
C6H6S2
MFCD00142541
1,4-benzenedithiol, dithiohydroquinone, 1,4-dimercaptobenzene, 4-mercaptothiophenol, 1,4-benzene dithiol, pubchem6866, acmc-1b45q, benzene-1,4-dithiol gc
benzene-1,4-dithiol

Specifications

624-39-5
White-Yellow
MFCD00142541
1,4-benzenedithiol, dithiohydroquinone, 1,4-dimercaptobenzene, 4-mercaptothiophenol, 1,4-benzene dithiol, pubchem6866, acmc-1b45q, benzene-1,4-dithiol gc
SC1=CC=C(S)C=C1
142.23
142.23
Crystalline Powder
97°C
C6H6S2
1 g
WYLQRHZSKIDFEP-UHFFFAOYSA-N
benzene-1,4-dithiol
4691729
≥98.0% (GC)
1,4-Benzenedithiol
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents