missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenedithiol 98.0+%, TCI America™

Catalog No. B15051G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15051G 1 g
B15055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15051G Supplier TCI America Supplier No. B15051G

Chemical Identifiers

CAS 624-39-5
Molecular Formula C6H6S2
Molecular Weight (g/mol) 142.23
MDL Number MFCD00142541
InChI Key WYLQRHZSKIDFEP-UHFFFAOYSA-N
Synonym 1,4-benzenedithiol, dithiohydroquinone, 1,4-dimercaptobenzene, 4-mercaptothiophenol, 1,4-benzene dithiol, pubchem6866, acmc-1b45q, benzene-1,4-dithiol gc
PubChem CID 4691729
IUPAC Name benzene-1,4-dithiol
SMILES SC1=CC=C(S)C=C1

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 1 g
Formula Weight 142.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Benzenedithiol