missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Benzenedimethanol 99.0+%, TCI America™
$50.05 - $354.23
Chemical Identifiers
| CAS | 589-29-7 |
|---|---|
| Molecular Formula | C8H10O2 |
| Molecular Weight (g/mol) | 138.166 |
| MDL Number | MFCD00004665 |
| InChI Key | BWVAOONFBYYRHY-UHFFFAOYSA-N |
| Synonym | 1,4-benzenedimethanol, p-xylylene glycol, p-xylene glycol, terephthalyl alcohol, 4-hydroxymethyl phenyl methanol, 1,4-xylylene glycol, 1,4-phenylenedimethanol, p-benzenedimethanol, p-xylylenediol, 1,4-dimethylolbenzene |
| PubChem CID | 11506 |
| IUPAC Name | [4-(hydroxymethyl)phenyl]methanol |
| SMILES | C1=CC(=CC=C1CO)CO |
Chemical Identifiers
| 589-29-7 | |
| 138.166 | |
| BWVAOONFBYYRHY-UHFFFAOYSA-N | |
| 11506 | |
| C1=CC(=CC=C1CO)CO |
| C8H10O2 | |
| MFCD00004665 | |
| 1,4-benzenedimethanol, p-xylylene glycol, p-xylene glycol, terephthalyl alcohol, 4-hydroxymethyl phenyl methanol, 1,4-xylylene glycol, 1,4-phenylenedimethanol, p-benzenedimethanol, p-xylylenediol, 1,4-dimethylolbenzene | |
| [4-(hydroxymethyl)phenyl]methanol |
Specifications
| 589-29-7 | |
| White-Yellow | |
| C8H10O2 | |
| 25 g | |
| BWVAOONFBYYRHY-UHFFFAOYSA-N | |
| [4-(hydroxymethyl)phenyl]methanol | |
| 11506 | |
| ≥99.0% (GC) | |
| 1,4-Benzenedimethanol |
| 118°C | |
| 143°C | |
| MFCD00004665 | |
| 1,4-benzenedimethanol, p-xylylene glycol, p-xylene glycol, terephthalyl alcohol, 4-hydroxymethyl phenyl methanol, 1,4-xylylene glycol, 1,4-phenylenedimethanol, p-benzenedimethanol, p-xylylenediol, 1,4-dimethylolbenzene | |
| C1=CC(=CC=C1CO)CO | |
| 138.166 | |
| 138.17 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1055
TSCA : Yes