missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenedimethanol 99.0+%, TCI America™

Catalog No. D060525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D060525G 25 g
D0605100G 100 g
D0605500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D060525G Supplier TCI America Supplier No. D060525G
Only null left

Chemical Identifiers

CAS 589-29-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00004665
InChI Key BWVAOONFBYYRHY-UHFFFAOYSA-N
Synonym 1,4-benzenedimethanol, p-xylylene glycol, p-xylene glycol, terephthalyl alcohol, 4-hydroxymethyl phenyl methanol, 1,4-xylylene glycol, 1,4-phenylenedimethanol, p-benzenedimethanol, p-xylylenediol, 1,4-dimethylolbenzene
PubChem CID 11506
IUPAC Name [4-(hydroxymethyl)phenyl]methanol
SMILES C1=CC(=CC=C1CO)CO

Specifications

Melting Point 118°C
Color White-Yellow
Boiling Point 143°C
Quantity 25 g
Formula Weight 138.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Benzenedimethanol
EINECSNumber (3)-1055
TSCA Yes