missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenedimethanol, 99%

Catalog No. AC105280500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105280500 50 g Glass bottle
AC105280100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105280500 Supplier Thermo Scientific Chemicals Supplier No. 105280500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 589-29-7 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 589-29-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004665
InChI Key BWVAOONFBYYRHY-UHFFFAOYSA-N
Synonym 1,4-benzenedimethanol, p-xylylene glycol, p-xylene glycol, terephthalyl alcohol, 4-hydroxymethyl phenyl methanol, 1,4-xylylene glycol, 1,4-phenylenedimethanol, p-benzenedimethanol, p-xylylenediol, 1,4-dimethylolbenzene
PubChem CID 11506
IUPAC Name [4-(hydroxymethyl)phenyl]methanol
SMILES C1=CC(=CC=C1CO)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 116.0°C to 119.0°C
Color White to Yellow
Boiling Point 138.0°C to 143.0°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H4(CH2OH)2
Quantity 50 g
Beilstein 06, 919
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,very soluble in ether & toluene
Formula Weight 138.17
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 1, 4-Benzenedimethanol, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-641-2
TSCA TSCA

RUO – Research Use Only