missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenedimethanol, 99%

Catalog No. AAA1440506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1440506 5 g
AAA1440514 25 g
AAA1440522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1440506 Supplier Thermo Scientific Chemicals Supplier No. A1440506
Only null left

CAS: 589-29-7 | C8H10O2 | 138.166 g/mol

1,4-Benzenedimethanol is used as an organic synthesis intermediate. It is also used for preparation of soluble polyphenylene and p-tolyl-methanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Benzenedimethanol is used as an organic synthesis intermediate. It is also used for preparation of soluble polyphenylene and p-tolyl-methanol.

Solubility
Soluble in ether, toluene, methanol and hot water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.

Chemical Identifiers

CAS 589-29-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00004665
InChI Key BWVAOONFBYYRHY-UHFFFAOYSA-N
Synonym 1,4-benzenedimethanol, p-xylylene glycol, p-xylene glycol, terephthalyl alcohol, 4-hydroxymethyl phenyl methanol, 1,4-xylylene glycol, 1,4-phenylenedimethanol, p-benzenedimethanol, p-xylylenediol, 1,4-dimethylolbenzene
PubChem CID 11506
IUPAC Name [4-(hydroxymethyl)phenyl]methanol
SMILES C1=CC(=CC=C1CO)CO

Specifications

Melting Point 117°C to 121°C
Boiling Point 138°C to 143°C (1 mmHg)
Quantity 5 g
Beilstein 2042077
Solubility Information Soluble in ether,toluene,methanol and hot water.
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material 1,4-Benzenedimethanol
EINECSNumber 209-641-2
TSCA Yes

RUO – Research Use Only