missing translation for 'onlineSavingsMsg'
Learn More

1,4-Anthraquinone, 94%

CAS: 635-12-1 | C14H8O2 | 208.216 g/mol

$208.30 - $752.01

Chemical Identifiers

CAS 635-12-1
Molecular Formula C14H8O2
Molecular Weight (g/mol) 208.216
MDL Number MFCD00013724
InChI Key LSOTZYUVGZKSHR-UHFFFAOYSA-N
Synonym 1,4-anthraquinone, 1,4-anthracenedione, 1,4-dioxoanthracene, 1,4-anthracenequinone, acmc-1b3md, 1,4-dihydroanthracene-1,4-dione
PubChem CID 69457
IUPAC Name anthracene-1,4-dione
SMILES C1=CC=C2C=C3C(=O)C=CC(=O)C3=CC2=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0057306
View Documents
Thermo Scientific Chemicals
L0057306
5 g
Each for $208.30
Only null left
 
AAL0057314
View Documents
Thermo Scientific Chemicals
L0057314
25 g
Each for $752.01
Only null left
 
Description

Description

1,4-Anthraquinone is used in the preparation of 1-methyl-1,4,4a,12a-tetrahydronaphthacene-5,12-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Anthraquinone is used in the preparation of 1-methyl-1,4,4a,12a-tetrahydronaphthacene-5,12-dione.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

635-12-1
208.216
LSOTZYUVGZKSHR-UHFFFAOYSA-N
69457
C1=CC=C2C=C3C(=O)C=CC(=O)C3=CC2=C1
C14H8O2
MFCD00013724
1,4-anthraquinone, 1,4-anthracenedione, 1,4-dioxoanthracene, 1,4-anthracenequinone, acmc-1b3md, 1,4-dihydroanthracene-1,4-dione
anthracene-1,4-dione

Specifications

635-12-1
C14H8O2
5 g
1,4-anthraquinone, 1,4-anthracenedione, 1,4-dioxoanthracene, 1,4-anthracenequinone, acmc-1b3md, 1,4-dihydroanthracene-1,4-dione
LSOTZYUVGZKSHR-UHFFFAOYSA-N
anthracene-1,4-dione
69457
94%
∽218°C (decomposition)
MFCD00013724
1871197
Insoluble in water.
C1=CC=C2C=C3C(=O)C=CC(=O)C3=CC2=C1
208.216
208.22
1,4-Anthraquinone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-228-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only