Learn More
1,4-Anthraquinone, 94%
CAS: 635-12-1 | C14H8O2 | 208.216 g/mol
Supplier: Thermo Scientific Chemicals L0057306
Description
1,4-Anthraquinone is used in the preparation of 1-methyl-1,4,4a,12a-tetrahydronaphthacene-5,12-dione.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Anthraquinone is used in the preparation of 1-methyl-1,4,4a,12a-tetrahydronaphthacene-5,12-dione.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
| 1,4-Anthraquinone | |
| ∽218°C (decomposition) | |
| MFCD00013724 | |
| 1871197 | |
| Insoluble in water. | |
| C1=CC=C2C=C3C(=O)C=CC(=O)C3=CC2=C1 | |
| 208.216 | |
| 208.22 |
| 635-12-1 | |
| C14H8O2 | |
| 5 g | |
| 1,4-anthraquinone, 1,4-anthracenedione, 1,4-dioxoanthracene, 1,4-anthracenequinone, acmc-1b3md, 1,4-dihydroanthracene-1,4-dione | |
| LSOTZYUVGZKSHR-UHFFFAOYSA-N | |
| anthracene-1,4-dione | |
| 69457 | |
| 94% |
Chemical Identifiers
| 635-12-1 | |
| 208.216 | |
| LSOTZYUVGZKSHR-UHFFFAOYSA-N | |
| 69457 | |
| C1=CC=C2C=C3C(=O)C=CC(=O)C3=CC2=C1 |
| C14H8O2 | |
| MFCD00013724 | |
| 1,4-anthraquinone, 1,4-anthracenedione, 1,4-dioxoanthracene, 1,4-anthracenequinone, acmc-1b3md, 1,4-dihydroanthracene-1,4-dione | |
| anthracene-1,4-dione |
Safety and Handling
EINECSNumber : 211-228-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only