missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4,8,11-Tetrathiacyclotetradecane 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 123°C |
| Color | White-Yellow |
| Quantity | 200 mg |
| Formula Weight | 268.51 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,4,8,11-Tetrathiacyclotetradecane |
Safety and Handling
| TSCA | No |