missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4,7-Trimethyl-1,4,7-triazacyclononane (stabilized with NaHCO3) 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| UN Number | 3267 |
| Formula Weight | 171.29 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,4,7-Trimethyl-1,4,7-triazacyclononane (stabilized with NaHCO3) |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |