CAS RN 96556-05-7
CAS RN 96556-05-7
IUPAC Name:
1,4,7-trimethyl-1,4,7-triazonane
Synonyms:
1,4,7-trimethyl-1,4,7-triazonane
Molecular Weight (g/mol):
171.29
Molecular Formula:
C9H21N3
InChi Key:
WLDGDTPNAKWAIR-UHFFFAOYSA-N
SMILES:
CN1CCN(C)CCN(C)CC1
1
–
1
of
1
results
1
1,4,7-Trimethyl-1,4,7-triazacyclononane (stabilized with NaHCO3) 98.0+%, TCI America™
CAS: 96556-05-7 Molecular Formula: C9H21N3 Molecular Weight (g/mol): 171.288 InChI Key: WLDGDTPNAKWAIR-UHFFFAOYSA-N Synonym: Octahydro-1,4,7-trimethyl-1H-1,4,7-triazonine PubChem CID: 546957 IUPAC Name: 1,4,7-trimethyl-1,4,7-triazonane SMILES: CN1CCN(CCN(CC1)C)C