missing translation for 'onlineSavingsMsg'
Learn More

1,4,7-Trimethyl-1,4,7-triazacyclononane (stabilized with NaHCO3) 98.0+%, TCI America™

Catalog No. T18791G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18791G 1 g
T18795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18791G Supplier TCI America Supplier No. T18791G

Chemical Identifiers

CAS 96556-05-7
Molecular Formula C9H21N3
Molecular Weight (g/mol) 171.288
InChI Key WLDGDTPNAKWAIR-UHFFFAOYSA-N
Synonym Octahydro-1,4,7-trimethyl-1H-1,4,7-triazonine
PubChem CID 546957
IUPAC Name 1,4,7-trimethyl-1,4,7-triazonane
SMILES CN1CCN(CCN(CC1)C)C

Specifications

Color Yellow
Quantity 1 g
UN Number 3267
Formula Weight 171.29
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,4,7-Trimethyl-1,4,7-triazacyclononane (stabilized with NaHCO3)
TSCA No
Recommended Storage Refrigerator