missing translation for 'onlineSavingsMsg'
Learn More

1,4,7-Triazacyclononane Trihydrochloride 98.0+%, TCI America™

Catalog No. p-7136865
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
T16001G 1 g
T16005G 5 g
2 options

Catalog No. T16001G

Supplier: TCI America T16001G

Only null left

Chemical Identifiers

CAS 58966-93-1
Molecular Formula C6H18N3
Molecular Weight (g/mol) 132.23
MDL Number MFCD00074887
InChI Key ITWBWJFEJCHKSN-UHFFFAOYSA-Q
Synonym 1,4,7-triazonane trihydrochloride, 1,4,7-triazacyclononane trihydrochloride, 1,4,7-triazacyclononane tri hydrochloride, octahydro-1h-1,4,7-triazonine trihydrochloride, 1h-1,4,7-triazonine, octahydro-, trihydrochloride, pubchem21332, 1,4,7-triazacyclononanetrihydrochloride, acmc-1au73, c6h15n3.3hcl, ksc911s7j
PubChem CID 2724990
IUPAC Name 1,4,7-triazonane-1,4,7-triium
SMILES C1C[NH2+]CC[NH2+]CC[NH2+]1

Specifications

Color White
Quantity 1 g
UN Number 1759
Formula Weight 238.58
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,7-Triazacyclononane Trihydrochloride