accessibility menu, dialog, popup

UserName

NH2NH2H2N

CAS RN 58966-93-1

IUPAC Name: 1,4,7-triazonane-1,4,7-triium
Synonyms: 1,4,7-triazonane-1,4,7-triium
Molecular Weight (g/mol): 132.23
Molecular Formula: C6H18N3
InChi Key: ITWBWJFEJCHKSN-UHFFFAOYSA-Q
SMILES: C1C[NH2+]CC[NH2+]CC[NH2+]1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)