missing translation for 'onlineSavingsMsg'
Learn More

1,4,7-Triazacyclononane trihydrochloride, 97%

Catalog No. p-202153
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294315000 500 mg Glass bottle
AC294310025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC294315000 Supplier Thermo Scientific Chemicals Supplier No. 294315000

CAS: 58966-93-1 | C6H18N3 | 132.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 58966-93-1
Molecular Formula C6H18N3
Molecular Weight (g/mol) 132.23
MDL Number MFCD00074887
InChI Key ITWBWJFEJCHKSN-UHFFFAOYSA-Q
Synonym 1,4,7-triazonane trihydrochloride, 1,4,7-triazacyclononane trihydrochloride, 1,4,7-triazacyclononane tri hydrochloride, octahydro-1h-1,4,7-triazonine trihydrochloride, 1h-1,4,7-triazonine, octahydro-, trihydrochloride, pubchem21332, 1,4,7-triazacyclononanetrihydrochloride, acmc-1au73, c6h15n3.3hcl, ksc911s7j
PubChem CID 2724990
SMILES C1C[NH2+]CC[NH2+]CC[NH2+]1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 285.0°C to 290.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 500 mg
IUPAC Name 1,4,7-triazonane-1,4,7-triium
Formula Weight 238.59
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1, 4, 7-Triazacyclononane trihydrochloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
TSCA TSCA

RUO – Research Use Only