missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4,7-Triazacyclononane 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 45°C |
| Color | White-Yellow |
| Boiling Point | 113°C |
| Quantity | 200 mg |
| UN Number | 3259 |
| Formula Weight | 129.21 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,4,7-Triazacyclononane |
Safety and Handling
| TSCA | No |