CAS RN 4730-54-5
IUPAC Name:
1,4,7-triazonane
Synonyms:
1,4,7-triazacyclononane tacn
Molecular Weight (g/mol):
129.21
Molecular Formula:
C6H15N3
InChi Key:
ITWBWJFEJCHKSN-UHFFFAOYSA-N
SMILES:
C1CNCCNCCN1
1
–
1
of
1
results
1
1,4,7-Triazacyclononane 98.0+%, TCI America™
CAS: 4730-54-5 Molecular Formula: C6H15N3 Molecular Weight (g/mol): 129.207 InChI Key: ITWBWJFEJCHKSN-UHFFFAOYSA-N Synonym: Octahydro-1H-1,4,7-triazonine PubChem CID: 188318 ChEBI: CHEBI:37405 IUPAC Name: 1,4,7-triazonane SMILES: C1CNCCNCCN1