missing translation for 'onlineSavingsMsg'
Learn More

1,4,7,10-Tetraazacyclododecane Tetrahydrochloride 98.0+%, TCI America™

Catalog No. T14261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14261G 1 g
T14265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14261G Supplier TCI America Supplier No. T14261G
Only null left

Chemical Identifiers

CAS 10045-25-7
Molecular Formula C8H24Cl4N4
Molecular Weight (g/mol) 318.108
MDL Number MFCD00068484
InChI Key SBWLCGZEBQGYRP-UHFFFAOYSA-N
Synonym 1,4,7,10-tetraazacyclododecane tetrahydrochloride, cyclen tetrahydrochloride, cyclen-4hcl, unii-51830y7tix, tetraaza-12-crown-4 tetrahydrochloride, 1,4,7,10-tetraazacyclododecane, tetrahydrochloride, 1,4,7,10-tetraazacyclododecane, hydrochloride 1:4, 1,4,7,10-tetraazacyclododecanetetrahydrochloride, cyclen tetrahydrochloride mi, 1,4,7,10-tetraazacyclododecane, chloride, chloride, chloride, chloride
PubChem CID 11652630
IUPAC Name 1,4,7,10-tetrazacyclododecane;tetrahydrochloride
SMILES C1CNCCNCCNCCN1.Cl.Cl.Cl.Cl

Specifications

Melting Point 280°C
Color White-Yellow
Quantity 1 g
Formula Weight 318.11
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,7,10-Tetraazacyclododecane Tetrahydrochloride