CAS RN 10045-25-7
IUPAC Name:
1,4,7,10-tetraazacyclododecane tetrahydrochloride
Synonyms:
cyclen tetrahydrochloride
Molecular Weight (g/mol):
318.11
Molecular Formula:
C8H24Cl4N4
InChi Key:
SBWLCGZEBQGYRP-UHFFFAOYSA-N
SMILES:
Cl.Cl.Cl.Cl.C1CNCCNCCNCCN1
1
–
1
of
1
results
1
1,4,7,10-Tetraazacyclododecane Tetrahydrochloride 98.0+%, TCI America™
CAS: 10045-25-7 Molecular Formula: C8H24Cl4N4 Molecular Weight (g/mol): 318.108 MDL Number: MFCD00068484 InChI Key: SBWLCGZEBQGYRP-UHFFFAOYSA-N Synonym: 1,4,7,10-tetraazacyclododecane tetrahydrochloride,cyclen tetrahydrochloride,cyclen-4hcl,unii-51830y7tix,tetraaza-12-crown-4 tetrahydrochloride,1,4,7,10-tetraazacyclododecane, tetrahydrochloride,1,4,7,10-tetraazacyclododecane, hydrochloride 1:4,1,4,7,10-tetraazacyclododecanetetrahydrochloride,cyclen tetrahydrochloride mi,1,4,7,10-tetraazacyclododecane, chloride, chloride, chloride, chloride PubChem CID: 11652630 IUPAC Name: 1,4,7,10-tetrazacyclododecane;tetrahydrochloride SMILES: C1CNCCNCCNCCN1.Cl.Cl.Cl.Cl