missing translation for 'onlineSavingsMsg'
Learn More

1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic Acid 97.0+%, TCI America™

Catalog No. T1875200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1875200MG 200 mg
T18751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1875200MG Supplier TCI America Supplier No. T1875200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60239-18-1
Molecular Formula C16H28N4O8
Molecular Weight (g/mol) 404.42
MDL Number MFCD00068657
InChI Key WDLRUFUQRNWCPK-UHFFFAOYSA-N
Synonym DOTA
PubChem CID 121841
ChEBI CHEBI:61028
IUPAC Name 2-[4,7,10-tris(carboxymethyl)-1,4,7,10-tetrazacyclododec-1-yl]acetic acid
SMILES C1CN(CCN(CCN(CCN1CC(=O)O)CC(=O)O)CC(=O)O)CC(=O)O

Specifications

Melting Point 267°C
Color White-Yellow
Quantity 200 mg
Formula Weight 404.42
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic Acid