missing translation for 'onlineSavingsMsg'
Learn More

1,4,5,8-Tetrachloroanthraquinone 95.0+%, TCI America™

Catalog No. p-7137683
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33005G 5 g
T330025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33005G Supplier TCI America Supplier No. T33005G

Chemical Identifiers

CAS 81-58-3
Molecular Formula C14H4Cl4O2
Molecular Weight (g/mol) 345.98
MDL Number MFCD00035801
InChI Key DUJPMUKIEFLXRE-UHFFFAOYSA-N
PubChem CID 66484
IUPAC Name 1,4,5,8-tetrachloro-9,10-dihydroanthracene-9,10-dione
SMILES ClC1=CC=C(Cl)C2=C1C(=O)C1=C(Cl)C=CC(Cl)=C1C2=O

Specifications

Melting Point 342°C
Color Yellow
Quantity 5 g
Formula Weight 345.98
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,5,8-Tetrachloroanthraquinone