accessibility menu, dialog, popup

UserName

CAS RN 81-58-3

IUPAC Name: 1,4,5,8-tetrachloro-9,10-dihydroanthracene-9,10-dione
Synonyms: 1,4,5,8-tetrachloroanthracene-9,10-dione
Molecular Weight (g/mol): 345.98
Molecular Formula: C14H4Cl4O2
InChi Key: DUJPMUKIEFLXRE-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)C2=C1C(=O)C1=C(Cl)C=CC(Cl)=C1C2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results