missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(1H-Indol-3-yl)piperidino]ethan-1-one, 97%, Thermo Scientific™

Chemical Identifiers

CAS 30030-83-2
Molecular Formula C15H18N2O
Molecular Weight (g/mol) 242.322
MDL Number MFCD01764639
InChI Key PRFIVCWVSORYRV-UHFFFAOYSA-N
Synonym 1-4-1h-indol-3-yl piperidino ethan-1-one, 1-4-1h-indol-3-yl piperidin-1-yl ethanone, maybridge3_005459, 3-n-acetyl-4-piperidyl indole, 1-acetyl-4-indol-3-ylpiperidine, 3-1-acetyl-4-piperidyl indole, 1-acetyl-4-1h-indol-3-yl piperidine, 3-1-acetylpiperidin-4-yl-1h-indole, 1-4-1h-indol-3-yl-piperidin-1-yl-ethanone
PubChem CID 2728359
IUPAC Name 1-[4-(1H-indol-3-yl)piperidin-1-yl]ethanone
SMILES CC(=O)N1CCC(CC1)C2=CNC3=CC=CC=C32
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RH01995DA
SDS View Documents
Thermo Scientific Chemicals
RH01995DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

30030-83-2
242.322
PRFIVCWVSORYRV-UHFFFAOYSA-N
2728359
CC(=O)N1CCC(CC1)C2=CNC3=CC=CC=C32
C15H18N2O
MFCD01764639
1-4-1h-indol-3-yl piperidino ethan-1-one, 1-4-1h-indol-3-yl piperidin-1-yl ethanone, maybridge3_005459, 3-n-acetyl-4-piperidyl indole, 1-acetyl-4-indol-3-ylpiperidine, 3-1-acetyl-4-piperidyl indole, 1-acetyl-4-1h-indol-3-yl piperidine, 3-1-acetylpiperidin-4-yl-1h-indole, 1-4-1h-indol-3-yl-piperidin-1-yl-ethanone
1-[4-(1H-indol-3-yl)piperidin-1-yl]ethanone

Specifications

30030-83-2
97%
C15H18N2O
1g
PRFIVCWVSORYRV-UHFFFAOYSA-N
1-[4-(1H-indol-3-yl)piperidin-1-yl]ethanone
2728359
97%
97.0
Amber Glass Bottle
MFCD01764639
1-4-1h-indol-3-yl piperidino ethan-1-one, 1-4-1h-indol-3-yl piperidin-1-yl ethanone, maybridge3_005459, 3-n-acetyl-4-piperidyl indole, 1-acetyl-4-indol-3-ylpiperidine, 3-1-acetyl-4-piperidyl indole, 1-acetyl-4-1h-indol-3-yl piperidine, 3-1-acetylpiperidin-4-yl-1h-indole, 1-4-1h-indol-3-yl-piperidin-1-yl-ethanone
CC(=O)N1CCC(CC1)C2=CNC3=CC=CC=C32
242.322
242.32
1-[4-(1H-Indol-3-yl)piperidino]ethan-1-one
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'