accessibility menu, dialog, popup

UserName

CAS RN 30030-83-2

CH3ONHN

CAS RN 30030-83-2

IUPAC Name: 1-[4-(1H-indol-3-yl)piperidin-1-yl]ethan-1-one
Synonyms: 1-[4-(1H-indol-3-yl)piperidin-1-yl]ethanone
Molecular Weight (g/mol): 242.32
Molecular Formula: C15H18N2O
InChi Key: PRFIVCWVSORYRV-UHFFFAOYSA-N
SMILES: CC(=O)N1CCC(CC1)C1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)