missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[3-(Trimethoxysilyl)propyl]urea 94.0+%, TCI America™
$68.97 - $518.21
Chemical Identifiers
| CAS | 23843-64-3 |
|---|---|
| Molecular Formula | C7H18N2O4Si |
| Molecular Weight (g/mol) | 222.32 |
| MDL Number | MFCD00216654 |
| InChI Key | LVACOMKKELLCHJ-UHFFFAOYSA-N |
| Synonym | (3-Ureidopropyl)trimethoxysilane |
| PubChem CID | 90280 |
| IUPAC Name | [3-(trimethoxysilyl)propyl]urea |
| SMILES | CO[Si](CCCNC(N)=O)(OC)OC |
Chemical Identifiers
| 23843-64-3 | |
| 222.32 | |
| LVACOMKKELLCHJ-UHFFFAOYSA-N | |
| 90280 | |
| CO[Si](CCCNC(N)=O)(OC)OC |
| C7H18N2O4Si | |
| MFCD00216654 | |
| (3-Ureidopropyl)trimethoxysilane | |
| [3-(trimethoxysilyl)propyl]urea |
Specifications
| 23843-64-3 | |
| C7H18N2O4Si | |
| 25 g | |
| LVACOMKKELLCHJ-UHFFFAOYSA-N | |
| [3-(trimethoxysilyl)propyl]urea | |
| 90280 | |
| ≥94.0% (N) | |
| 1-[3-(Trimethoxysilyl)propyl]urea |
| Yellow | |
| MFCD00216654 | |
| (3-Ureidopropyl)trimethoxysilane | |
| CO[Si](CCCNC(N)=O)(OC)OC | |
| 222.32 | |
| 222.32 | |
| Liquid |
Safety and Handling
TSCA : Yes