accessibility menu, dialog, popup

UserName

CH3OSiNHH2NOOH3COH3C

CAS RN 23843-64-3

IUPAC Name: [3-(trimethoxysilyl)propyl]urea
Synonyms: 3-(trimethoxysilyl)propylurea
Molecular Weight (g/mol): 222.32
Molecular Formula: C7H18N2O4Si
InChi Key: LVACOMKKELLCHJ-UHFFFAOYSA-N
SMILES: CO[Si](CCCNC(N)=O)(OC)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results