missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Pyridyl)ethylamine, 96%

Catalog No. p-7028650
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26047MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH26047MD Supplier Thermo Scientific Chemicals Supplier No. H26047MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56129-55-6 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56129-55-6
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD01691641,MFCD09256803
InChI Key IQVQNBXPYJGNEA-UHFFFAOYNA-N
Synonym 1-pyridin-3-yl-ethylamine, 1-3-pyridyl ethylamine, 1-pyridin-3-yl ethanamine, 3-1-aminoethyl pyridine, 1-3-pyridinyl ethanamine, 1-pyridin-3-yl ethan-1-amine, alpha-methyl-3-pyridinemethylamine, 3-pyridinemethylamine, alpha-methyl, 1-pyridin-3-ylethyl amine, 1-3-pyridyl ethanamine
PubChem CID 2771688
IUPAC Name 1-pyridin-3-ylethanamine
SMILES CC(N)C1=CC=CN=C1

Specifications

Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 122.17
Percent Purity 96%
Chemical Name or Material 1-(3-Pyridyl)ethylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only