CAS RN 56129-55-6
IUPAC Name:
1-(pyridin-3-yl)ethan-1-amine
Synonyms:
1-(pyridin-3-yl)ethanamine
Molecular Weight (g/mol):
122.17
Molecular Formula:
C7H10N2
InChi Key:
IQVQNBXPYJGNEA-UHFFFAOYNA-N
SMILES:
CC(N)C1=CC=CN=C1
1
–
1
of
1
results
1
Promotions Available
1-(3-Pyridyl)ethylamine, 96%
1-(3-Pyridyl)ethylamine, 96%
CAS: 56129-55-6 Molecular Formula: C7H10N2 Molecular Weight (g/mol): 122.17 MDL Number: MFCD01691641,MFCD09256803 InChI Key: IQVQNBXPYJGNEA-UHFFFAOYNA-N Synonym: 1-pyridin-3-yl-ethylamine,1-3-pyridyl ethylamine,1-pyridin-3-yl ethanamine,3-1-aminoethyl pyridine,1-3-pyridinyl ethanamine,1-pyridin-3-yl ethan-1-amine,alpha-methyl-3-pyridinemethylamine,3-pyridinemethylamine, alpha-methyl,1-pyridin-3-ylethyl amine,1-3-pyridyl ethanamine PubChem CID: 2771688 IUPAC Name: 1-pyridin-3-ylethanamine SMILES: CC(N)C1=CC=CN=C1