missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanesultone 99.0+%, TCI America™

Catalog No. P0324500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0324500G 500 g
P032425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0324500G Supplier TCI America Supplier No. P0324500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1120-71-4
Molecular Formula C3H6O3S
Molecular Weight (g/mol) 122.14
MDL Number MFCD00005355
InChI Key FSSPGSAQUIYDCN-UHFFFAOYSA-N
Synonym 1,3-propanesultone, propane sultone, 1,2-oxathiolane 2,2-dioxide, 1,3-propane sultone, 1,2-oxathiolane, 2,2-dioxide, propanesultone, rcra waste number u193, .gamma.-propane sultone, 3-hydroxy-1-propanesulfonic acid gamma-sultone, unii-l6ntk7vjx9
PubChem CID 14264
ChEBI CHEBI:82370
IUPAC Name 1,2λ⁶-oxathiolane-2,2-dione
SMILES O=S1(=O)CCCO1

Specifications

Color White-Yellow
Boiling Point 180°C
Quantity 500 g
UN Number 2811
Formula Weight 122.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,3-Propanesultone
EINECSNumber (5)-0530
RTECSNumber RP5425000
TSCA Yes