accessibility menu, dialog, popup

UserName

CAS RN 1120-71-4

IUPAC Name: 1,2λ⁶-oxathiolane-2,2-dione
Synonyms: 1,3-propanesultone 1,3-propane sultone 2,2-dioxide-1,2-oxathiolane
Molecular Weight (g/mol): 122.14
Molecular Formula: C3H6O3S
InChi Key: FSSPGSAQUIYDCN-UHFFFAOYSA-N
SMILES: O=S1(=O)CCCO1
Boiling Point 
  • (2)
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)