missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanesultone, 99%

Catalog No. AAA1192330
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1192330 250 g
AAA1192309 10 g
AAA1192318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1192330 Supplier Thermo Scientific Chemicals Supplier No. A1192330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1120-71-4 | C3H6O3S | 122.14 g/mol

1,3-Propane sultone is used as a chemical intermediate to introduce sulfopropyl groups into molecules and to confer water solubility and anionic character.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Propane sultone is used as a chemical intermediate to introduce sulfopropyl groups into molecules and to confer water solubility and anionic character.

Solubility
Soluble in water (100 mg/ml), chloroform, esters

Chemical Identifiers

CAS 1120-71-4
Molecular Formula C3H6O3S
Molecular Weight (g/mol) 122.14
MDL Number MFCD00005355
InChI Key FSSPGSAQUIYDCN-UHFFFAOYSA-N
Synonym 1,3-propanesultone, propane sultone, 1,2-oxathiolane 2,2-dioxide, 1,3-propane sultone, 1,2-oxathiolane, 2,2-dioxide, propanesultone, rcra waste number u193, .gamma.-propane sultone, 3-hydroxy-1-propanesulfonic acid gamma-sultone, unii-l6ntk7vjx9
PubChem CID 14264
ChEBI CHEBI:82370
SMILES O=S1(=O)CCCO1

Specifications

Melting Point 30°C to 33°C
Density 1.392
Boiling Point 179°C to 181°C (30 mmHg)
Flash Point >110°C (230°F)
Quantity 250 g
UN Number UN2811
Beilstein 109782
Sensitivity Moisture sensitive
Solubility Information Soluble in water (100mg/ml),chloroform,esters,methanol,and aromatic hydrocartions.
IUPAC Name oxathiolane 2,2-dioxide
Formula Weight 122.14
Percent Purity 99%
Chemical Name or Material 1,3-Propanesultone
Show More Show Less
Hazard Category H302+H312-H350
Hazard Statement GHS H Statement
H350-H302-H312;
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 214-317-9
RTECSNumber RP5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only