missing translation for 'onlineSavingsMsg'
Learn More

1,3-Phenylenediamine 98.0+%, TCI America™

Catalog No. p1911250g
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1911250G 250 g
P191125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1911250G Supplier TCI America Supplier No. P1911250G
Only null left

Chemical Identifiers

CAS 108-45-2
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00007799
InChI Key WZCQRUWWHSTZEM-UHFFFAOYSA-N
Synonym m-phenylenediamine, 1,3-phenylenediamine, 1,3-diaminobenzene, 1,3-benzenediamine, m-diaminobenzene, 3-aminoaniline, m-aminoaniline, meta-phenylenediamine, developer c, 3-phenylenediamine
PubChem CID 7935
ChEBI CHEBI:8092
IUPAC Name benzene-1,3-diamine
SMILES C1=CC(=CC(=C1)N)N

Specifications

Melting Point 65°C
Color White
Boiling Point 287°C
Quantity 250 g
UN Number 1673
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Phenylenediamine
EINECSNumber (3)-0185
RTECSNumber SS7700000
TSCA Yes