missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Phenylenediamine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 65°C |
| Boiling Point | 287°C |
| Quantity | 25 g |
| UN Number | 1673 |
| Formula Weight | 108.14 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,3-Phenylenediamine |
Safety and Handling
| EINECSNumber | (3)-0185 |
| RTECSNumber | SS7700000 |
| TSCA | Yes |