missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Phenylenediamine Dihydrochloride 99.0+%, TCI America™
$61.61 - $577.56
Chemical Identifiers
| CAS | 541-69-5 |
|---|---|
| Molecular Formula | C6H10Cl2N2 |
| Molecular Weight (g/mol) | 181.06 |
| MDL Number | MFCD00012975 |
| InChI Key | SVTOYMIYCMHPIV-UHFFFAOYSA-N |
| Synonym | benzene-1,3-diamine dihydrochloride, m-phenylenediamine dihydrochloride, 1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine dihydrochloride, 1,3-benzenediamine, dihydrochloride, m-phenylenediamine hydrochloride, unii-se5rlo93tg, 1,3-diaminobenzene dihydrochloride, m-aminoaniline dihydrochloride, 3-aminoaniline dihydrochloride |
| PubChem CID | 10941 |
| IUPAC Name | benzene-1,3-diamine;dihydrochloride |
| SMILES | C1=CC(=CC(=C1)N)N.Cl.Cl |
Chemical Identifiers
| 541-69-5 | |
| 181.06 | |
| SVTOYMIYCMHPIV-UHFFFAOYSA-N | |
| 10941 | |
| C1=CC(=CC(=C1)N)N.Cl.Cl |
| C6H10Cl2N2 | |
| MFCD00012975 | |
| benzene-1,3-diamine dihydrochloride, m-phenylenediamine dihydrochloride, 1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine dihydrochloride, 1,3-benzenediamine, dihydrochloride, m-phenylenediamine hydrochloride, unii-se5rlo93tg, 1,3-diaminobenzene dihydrochloride, m-aminoaniline dihydrochloride, 3-aminoaniline dihydrochloride | |
| benzene-1,3-diamine;dihydrochloride |
Specifications
| 541-69-5 | |
| C6H10Cl2N2 | |
| 25 g | |
| benzene-1,3-diamine dihydrochloride, m-phenylenediamine dihydrochloride, 1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine dihydrochloride, 1,3-benzenediamine, dihydrochloride, m-phenylenediamine hydrochloride, unii-se5rlo93tg, 1,3-diaminobenzene dihydrochloride, m-aminoaniline dihydrochloride, 3-aminoaniline dihydrochloride | |
| C1=CC(=CC(=C1)N)N.Cl.Cl | |
| 181.06 | |
| 181.06 | |
| Crystalline Powder |
| Purple-White | |
| MFCD00012975 | |
| 2811 | |
| SVTOYMIYCMHPIV-UHFFFAOYSA-N | |
| benzene-1,3-diamine;dihydrochloride | |
| 10941 | |
| ≥99.0% (T) | |
| 1,3-Phenylenediamine Dihydrochloride |
Safety and Handling
EINECSNumber : (3)-0185
RTECSNumber : SS9800000
TSCA : Yes