missing translation for 'onlineSavingsMsg'
Learn More

1,3-Phenylenediamine Dihydrochloride 99.0+%, TCI America™

Catalog No. p017225g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
P017225G 25 g
P0172500G 500 g
2 options

Catalog No. P017225G

Supplier: TCI America P017225G

Only null left

Chemical Identifiers

CAS 541-69-5
Molecular Formula C6H10Cl2N2
Molecular Weight (g/mol) 181.06
MDL Number MFCD00012975
InChI Key SVTOYMIYCMHPIV-UHFFFAOYSA-N
Synonym benzene-1,3-diamine dihydrochloride, m-phenylenediamine dihydrochloride, 1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine dihydrochloride, 1,3-benzenediamine, dihydrochloride, m-phenylenediamine hydrochloride, unii-se5rlo93tg, 1,3-diaminobenzene dihydrochloride, m-aminoaniline dihydrochloride, 3-aminoaniline dihydrochloride
PubChem CID 10941
IUPAC Name benzene-1,3-diamine;dihydrochloride
SMILES C1=CC(=CC(=C1)N)N.Cl.Cl

Specifications

Color Purple-White
Quantity 25 g
UN Number 2811
Formula Weight 181.06
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Phenylenediamine Dihydrochloride
EINECSNumber (3)-0185
RTECSNumber SS9800000
TSCA Yes