missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Hydroxypropyl)pyrrole 98.0+%, TCI America™

Catalog No. H08271G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H08271G Supplier TCI America Supplier No. H08271G
Only null left

Chemical Identifiers

CAS 50966-69-3
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.171
MDL Number MFCD00191453
InChI Key ORCXGLCMROHGOM-UHFFFAOYSA-N
PubChem CID 12794991
IUPAC Name 3-pyrrol-1-ylpropan-1-ol
SMILES C1=CN(C=C1)CCCO

Specifications

Color Yellow
Boiling Point 231°C
Quantity 1 g
Formula Weight 125.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(3-Hydroxypropyl)pyrrole
TSCA No
Recommended Storage Refrigerator