accessibility menu, dialog, popup

UserName

NHO

CAS RN 50966-69-3

IUPAC Name: 3-(1H-pyrrol-1-yl)propan-1-ol
Synonyms: 3-(pyrrol-1-yl)propan-1-ol
Molecular Weight (g/mol): 125.17
Molecular Formula: C7H11NO
InChi Key: ORCXGLCMROHGOM-UHFFFAOYSA-N
SMILES: OCCCN1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results