missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Hydroxybenzyl)piperidine 98.0+%, TCI America™

Catalog No. H11835G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11835G 5 g
H118325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11835G Supplier TCI America Supplier No. H11835G
Only null left

Chemical Identifiers

CAS 73279-04-6
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD00800250
InChI Key ORGBERFQYFWYGX-UHFFFAOYSA-N
Synonym 3-1-piperidinylmethyl phenol, 3-piperidin-1-ylmethyl phenol, 3-1-piperidinomethyl phenol, 1-3-hydroxybenzyl piperidine, 3-piperidinomethyl phenol, 3-1-piperidylmethyl phenol, phenol, 3-1-piperidinylmethyl, 1-3-hydroxyphenylmethyl piperidine, pubchem6815, 3-piperidinomethylphenol
PubChem CID 826974
IUPAC Name 3-(piperidin-1-ylmethyl)phenol
SMILES C1CCN(CC1)CC2=CC(=CC=C2)O

Specifications

Melting Point 139°C
Color White-Yellow
Quantity 5 g
Formula Weight 191.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Hydroxybenzyl)piperidine